missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Adapters for Rotors

Adapters for Rotors are for use with a variety of Eppendorf™, Corning™ and Nalgene™ tubes and plates in different shapes and sizes.

Brand:  Eppendorf™ 5920742000

Additional Details : Weight : 0.90000kg

 View more versions of this product

Product Code. 15715882

  • £301.00 / Pack of 2
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Available for use with:
  • 2.6 to 8 mL, set of 2 pcs.
  • 250 mL flat, 175 to 225 mL conical, and Nunc™ 250 mL conical, set of 2 pcs.
    • Please note that these conical bottles need to be used together with a conical form insert provided by the manufacturer.
  • 49 x 2.6 to 8 mL, set of 2 pcs.
  • 5.5 to 12 mL, set of 2 pcs.
  • 500 mL Corning™, set of 2 pcs.
  • 500 mL Nalgene™, set of 2 pcs.
  • 50 mL, set of 2 pcs.
  • 5 mL FACS, set of 2 pcs.
  • 750 mL, set of 2 pcs.
  • 9 mL, set of 2 pcs.
  • Eppendorf Tubes™ 5.0 mL and 15 mL, set of 2 pcs.
  • MTP, DWP, PCR plates, set of 2 pcs.
Specifications

Specifications

Rotor Adapter
5 mL FACS
51
Round Bottom
Rotor S-4xUniversal-Large
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers