Learn More
Eppendorf™ Adapters for Rotors
Adapters for Rotors are for use with a variety of Eppendorf™, Corning™ and Nalgene™ tubes and plates in different shapes and sizes.
£286.00 - £301.00
Product Code | Brand | Capacity | Tube Type | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Tube Type | For Use With (Equipment) | Price | Quantity & Availability | |||
15758389
|
Eppendorf™
5920718002 |
2.6 to 8 mL | Round Bottom | Rotor S-4x1000, High-capacity Buckets |
£286.00
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15728399
|
Eppendorf™
5920739000 |
2.6 to 8 mL | Round Bottom | Rotor S-4xUniversal-Large |
£286.00
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15715882
|
Eppendorf™
5920742000 |
5 mL FACS | Round Bottom | Rotor S-4xUniversal-Large |
£301.00
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15718399
|
Eppendorf™
5920738003 |
5.5 to 12 mL | Round Bottom | Rotor S-4xUniversal-Large |
£286.00
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15778389
|
Eppendorf™
5920722000 |
9 mL | Round Bottom | Rotor S-4x1000, High-capacity Buckets |
£286.00
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15738399
|
Eppendorf™
5920740008 |
250 mL Flat, 175 to 225 mL Conical | Flat/Conical Bottom | Rotor S-4xUniversal-Large |
£286.00
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15748389
|
Eppendorf™
5920717006 |
250 mL Flat, 175 to 225 mL Conical, Nunc™ 250 mL Conical | Flat/Conical Bottom | Rotor S-4x1000, High-capacity Buckets |
£286.00
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15758399
|
Eppendorf™
5920744003 |
500 mL Corning™ Conical Bottle | Conical | Rotor S-4xUniversal-Large |
£286.00
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15768399
|
Eppendorf™
5920745000 |
500 mL Nalgene™ Flat Bottle | Flat Bottom | Rotor S-4xUniversal-Large |
£286.00
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15748399
|
Eppendorf™
5920741004 |
750 mL Bottle | Flat Bottom | Rotor S-4xUniversal-Large |
£286.00
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15638918
|
Eppendorf™
5920737007 |
6 x MTP, DWP, PCR Plates | Flat Bottom | S-4xUniversal-Large, Universal Buckets |
£286.00
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Descrizione
Available for use with:- 2.6 to 8 mL, set of 2 pcs.
- 250 mL flat, 175 to 225 mL conical, and Nunc™ 250 mL conical, set of 2 pcs.
- Please note that these conical bottles need to be used together with a conical form insert provided by the manufacturer.
- 49 x 2.6 to 8 mL, set of 2 pcs.
- 5.5 to 12 mL, set of 2 pcs.
- 500 mL Corning™, set of 2 pcs.
- 500 mL Nalgene™, set of 2 pcs.
- 50 mL, set of 2 pcs.
- 5 mL FACS, set of 2 pcs.
- 750 mL, set of 2 pcs.
- 9 mL, set of 2 pcs.
- Eppendorf Tubes™ 5.0 mL and 15 mL, set of 2 pcs.
- MTP, DWP, PCR plates, set of 2 pcs.
Specifica
Rotor Adapter |