Learn More
Eppendorf™ Adapter S-4x Universal Rotor Large
Facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for S-4xUniversal-Large rotor.
Brand:  Eppendorf™ 5920747002
Additional Details : Weight : 1.21500kg
 View more versions of this product
Product Code. 15788399
  • £301.00 / Pack of 2
In Stock
to see stock.
Description
Description
  • Suitable for S-4xUniversal-Large rotor buckets
  • Accommodates selected array of tubes, bottles, or cards
Specifications
Specifications
Microtube
11 mm
S-4xUniversal-Large
92
92 x 1.5/2.0 Microtubes
39 mm
92
2/Pk.