Learn More
Eppendorf™ Adapter For LC 481 TwinTec PCR Plate
GREENER_CHOICE
Product Code. 10319557 Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
For LC 481 TwinTec PCR Plate
Unit Size:
Each
Product Code. 10319557
Brand: Eppendorf™ 0030133412
Only null left
Add to basket
Add to basket
Eppendorf twin-tec PCR plates give you the advantages that come from combining two materials: the wells are manufactured from virgin polypropylene, the most successful material used for PCR tubes; polycarbonate, an especially rigid material, forms the plate deck and the skirt.
  • One-piece design achieved using twin-shot technology, an innovative injection moulding process
  • Minimal bending of skirts
  • Semi-skirted for labeling and bar-coding
  • Suitable for quantitative real-time PCR
  • Fits most Thermal Cyclers
  • 250μL max. well volume (when used with cap strips)
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
For Use With (Equipment) For LC 481 TwinTec PCR Plate
Quantity 1 Each