Learn More
Eppendorf™ Adapter for Rotor F-35-6-30
Adapter for Rotors designed for use with Eppendorf Centrifuges
Brand: Eppendorf™ 5427723005
Code : A0
Additional Details : Weight : 0.08600kg
Description
- Rotor F-35-6-30 holds 6 x 15/50mL conical or 12 × Vacutainer™ tubes
Specifications
Large Rotor Bore Adapter | |
1 x 100 to 125 mm Centriprep™ | |
Large rotor bore | |
6/Pk. |
Centriprep | |
Rotor F-35-6-30 | |
For Centriplus Centrifugal Filter Units |