Learn More
Honeywell™ Adaptec Safety Glasses, Regular
Product Code. 15876655
Change view
Click to view available options
Lens Tint:
Amber
Clear
Smoke
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Tint unitSize
15876655 Clear Each
15866655 Amber Each
15886655 Smoke Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15876655 Supplier Honeywell™ Supplier No. 1030749HS
Only null left
Add to basket
Add to basket
Designed to fit the widest range of workers, these safety glasses are stylish, comfortable, and adjustable, and feature excellent protection, optics, and Hydroshield anti-fog/anti-scratch coating.
  • Three-position nose bridge is simple to adjust and keeps eyes centered in the lens, where optical clarity is best
  • Workers' vision is less strained so they experience less eye fatigue, work more comfortably for longer periods and can more clearly see their work
  • Soft, flexible material allows each nose pad to independently conform to the wearer's face, delivering greater comfort, contact and stability
  • Adjustable lens angle improves safety and reduces fogging.
  • Each temple hinge ratchets to seven positions - 30 degrees of custom adjustability so lens is angled for both optimal eye protection and ideal space for air circulation
  • A proprietary temple design (patent pending) allows workers to adjust temple tension anytime, safely and without removing eyewear
  • Temple arm features a rigid insert that slides from front to back to control pressure against the head for a firmer or looser fit.
  • Available in regular, narrow, and wide designs
  • One style means simplicity of stocking and ordering as well as consistent eyewear usage by all workers in your facility. No longer is there a need to stock 3-5 different styles of eyewear from multiple manufacturers
  • Vented, cushioned frame for less fogging
  • The dual-action HydroShield anti-fog lens coating provides more than 20X longer lasting fog-free performance - even after extended wear and repeated cleanings and provides 2X greater scratch-resistance
TRUSTED_SUSTAINABILITY
Specifications
Lens Type Hydroshield coating
Lens Coating Anti-fog, Scratch-resistant
Description Dielectric
Certifications/Compliance (Europe) EC Category PPE: 2, QA: ISO 9001/2000, EC Certificate: C2317.1N, EC Attestation: C2317.1N
Lens Tint Clear
For Use With (Application) Building, construction, general industry, maintenance
Frame Color Red, Black
Lens Material Polycarbonate
Frame Material Polycarbonate