missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTS19 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | ADAMTS19 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ADAMTS19 Polyclonal specifically detects ADAMTS19 in Human samples. It is validated for Western Blot.Specifications
| ADAMTS19 | |
| Polyclonal | |
| Rabbit | |
| Q8TE59 | |
| 171019 | |
| IgG | |
| 100 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| A disintegrin and metalloproteinase with thrombospondin motifs 19, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 19, ADAM metallopeptidase with thrombospondin type 1 motif, 19, ADAM-TS 19, ADAM-TS19, ADAMTS-19, EC 3.4.24.-, FLJ16042 | |
| ADAMTS19 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title