missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTS19 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69170
This item is not returnable.
View return policy
Description
ADAMTS19 Polyclonal specifically detects ADAMTS19 in Human samples. It is validated for Western Blot.
Specifications
| ADAMTS19 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| A disintegrin and metalloproteinase with thrombospondin motifs 19, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 19, ADAM metallopeptidase with thrombospondin type 1 motif, 19, ADAM-TS 19, ADAM-TS19, ADAMTS-19, EC 3.4.24.-, FLJ16042 | |
| Rabbit | |
| 100 kDa | |
| 100 μL | |
| Primary | |
| Rat: 77%. | |
| Human, Rat, Bovine, Canine | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q8TE59 | |
| ADAMTS19 | |
| Synthetic peptides corresponding to ADAMTS19 (ADAM metallopeptidase with thrombospondin type 1 motif, 19) The peptide sequence was selected from the N terminal of ADAMTS19 (NP_598377). Peptide sequence MRLTHICCCCLLYQLGFLSNGIVSELQFAPDREEWEVVFPALWRREPVDP The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| 171019 | |
| Centrifuge vial prior to reconstitution. Add 50μL distilled water to a final antibody concentration of 1mg/mL. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction