missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ADAMDEC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | ADAMDEC1 |
|---|---|
| Dilution | Western Blot : 1:400, ELISA, Immunohistochemistry-Paraffin : 2-10 μg/mL |
| Applications | ELISA, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066881
|
Novus Biologicals™
NBP3-30959-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067209
|
Novus Biologicals™
NBP3-30959-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ADAMDEC1 Polyclonal antibody specifically detects ADAMDEC1 in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)Specifications
| ADAMDEC1 | |
| ELISA, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| Phosphate buffered saline | |
| 27299 | |
| IgG | |
| Antigen Affinity-purified |
| Western Blot : 1:400, ELISA, Immunohistochemistry-Paraffin : 2-10 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| A disintegrin and metalloproteinase domain-like protein decysin-1, ADAM DEC1, ADAM-like protein decysin-1, ADAM-like, decysin 1, decysin, disintegrin protease, EC 3.4.24.-, FLJ79219, M12.219 | |
| Synthetic peptide derived from the human ADAMDEC1 protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title