missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ADAMDEC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30959-25ug
This item is not returnable.
View return policy
Description
ADAMDEC1 Polyclonal antibody specifically detects ADAMDEC1 in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| ADAMDEC1 | |
| Polyclonal | |
| Western Blot : 1:400, ELISA, Immunohistochemistry-Paraffin : 2-10 μg/mL | |
| A disintegrin and metalloproteinase domain-like protein decysin-1, ADAM DEC1, ADAM-like protein decysin-1, ADAM-like, decysin 1, decysin, disintegrin protease, EC 3.4.24.-, FLJ79219, M12.219 | |
| Synthetic peptide derived from the human ADAMDEC1 protein. | |
| 25 μg | |
| Signal Transduction | |
| 27299 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction