missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADA2a Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80023
This item is not returnable.
View return policy
Description
ADA2a Polyclonal specifically detects ADA2a in Human samples. It is validated for Western Blot.
Specifications
| ADA2a | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ADA2AFLJ12705, ADA2-like protein, hADA2, TADA2LADA2, transcriptional adapter 2-alpha, Transcriptional adapter 2-like, transcriptional adaptor 2 (ADA2 homolog, yeast)-like, transcriptional adaptor 2 alpha, transcriptional adaptor 2A | |
| Rabbit | |
| 51 kDa | |
| 100 μL | |
| Cell Cycle and Replication, mTOR Pathway | |
| 6871 | |
| Human | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_001479 | |
| TADA2A | |
| Synthetic peptide directed towards the C terminal of human TADA2L. Peptide sequence RRQADIDSGLSPSIPMASNSGRRSAPPLNLTGLPGTEKLNEKEKELCQMV. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction