missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADA, 0.2M buffer soln., pH 6.0, Thermo Scientific™
£40.90 - £55.35
Chemical Identifiers
CAS | 7415-22-7 |
---|---|
Molecular Formula | C6H9N2NaO5 |
Molecular Weight (g/mol) | 212.137 |
MDL Number | MFCD00065483 |
InChI Key | QCGKKVWCABJQQI-UHFFFAOYSA-M |
PubChem CID | 23679048 |
IUPAC Name | sodium;2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetate |
SMILES | C(C(=O)N)N(CC(=O)O)CC(=O)[O-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15402899
|
Thermo Scientific Alfa Aesar
J60054.AE |
100 mL |
£40.90
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15412899
|
Thermo Scientific Alfa Aesar
J60054.AK |
250 mL | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7415-22-7 | |
212.137 | |
QCGKKVWCABJQQI-UHFFFAOYSA-M | |
sodium;2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetate |
C6H9N2NaO5 | |
MFCD00065483 | |
23679048 | |
C(C(=O)N)N(CC(=O)O)CC(=O)[O-].[Na+] |
Specifications
7415-22-7 | |
0.2 M | |
Liquid | |
MFCD00065483 | |
C(C(=O)N)N(CC(=O)O)CC(=O)[O-].[Na+] | |
212.137 | |
ADA buffer soln. |
Colorless | |
6.0 | |
C6H9N2NaO5 | |
QCGKKVWCABJQQI-UHFFFAOYSA-M | |
sodium;2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetate | |
23679048 |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Keep cold