missing translation for 'onlineSavingsMsg'
Learn More

ADA, 0.2M buffer soln., pH 6.0, Thermo Scientific™

Product Code. 15402899
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15402899 100 mL 100mL
15412899 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15402899 Supplier Thermo Scientific Alfa Aesar Supplier No. J60054.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7415-22-7
Molecular Formula C6H9N2NaO5
Molecular Weight (g/mol) 212.137
MDL Number MFCD00065483
InChI Key QCGKKVWCABJQQI-UHFFFAOYSA-M
PubChem CID 23679048
IUPAC Name sodium;2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetate
SMILES C(C(=O)N)N(CC(=O)O)CC(=O)[O-].[Na+]

Specifications

Color Colorless
Concentration 0.2 M
pH 6.0
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material ADA buffer soln.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.