missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACVRL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14745-1-AP-150UL
This item is not returnable.
View return policy
Description
The TGF-beta superfamily of ligands exerts its biological activity by binding to heteromeric receptor complexes two types (I and II) of the serine/threonine kinases. ALK1, like the other type I receptors, contains a cysteine rich domain with conserved cysteine spacing in the extracellular region and a glycine and serine rich domain preceding the kinase domain. The reduced human ALK1 monomer has a calculated molecular mass of approximately 37.3 kDa. Due to glycoslyation, the recombit human protein migrates as a 50-55 kDa protein in SDS-PAGE under reducing conditions. Human and mouse ALK1 share approximately 71% amino acid sequence identity in their extracellular domains. ALK1 is highly expressed in endothelial cells and other vascular tissues.Specifications
| ACVRL1 | |
| Polyclonal | |
| Unconjugated | |
| ACVRL1 | |
| Activin receptor like kinase 1, ACVRL1, ACVRLK1, ALK 1, ALK1, HHT, HHT2, ORW2, SKR3, TSR I | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 94 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P37023 | |
| ACVRL1 | |
| ACVRL1 Fusion Protein Ag6449 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction