missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTR3B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56602
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
ACTR3B Polyclonal specifically detects ACTR3B in Human samples. It is validated for Western Blot.
Spezifikation
| ACTR3B | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| actin-related protein 3B, actin-related protein 3-beta, actin-related protein Arp11, Actin-related protein ARP4, ARP11Actin-like protein 3B, ARP3 actin-related protein 3 homolog B (yeast), ARP3beta, ARP3-beta, ARP4, DKFZp686O24114 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 57180 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9P1U1 | |
| ACTR3B | |
| Synthetic peptides corresponding to ACTR3B(ARP3 actin-related protein 3 homolog B (yeast)) Antibody(against the N terminal of ACTR3B. Peptide sequence MAGSLPPCVVDCGTGYTKLGYAGNTEPQFIIPSCIAIRESAKVVDQAQRR. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Chicken: 100%; Canine: 100%; Guinea pig: 100%; Equine: 100%; Rat: 100%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur