missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTR10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£349.00
Specifications
| Antigen | ACTR10 |
|---|---|
| Concentration | 0.24 mg/mL |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16859584
|
Proteintech
20101-1-AP-20UL |
20 μL |
N/A
|
N/A | |||||
|
16849584
|
Proteintech
20101-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The function of this protein remains unknown.Specifications
| ACTR10 | |
| Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| Q9NZ32, Q9QZB7 | |
| 299121, 55860, 56444 | |
| ACTR10 Fusion Protein Ag13612 | |
| Primary | |
| -20°C | |
| ACTR10 |
| 0.24 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Actin related protein 10, Actin related protein 11, ACTR10, ACTR11, ARP11, HARP11 | |
| ACTR10 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title