missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTR10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20101-1-AP-20UL
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
| ACTR10 | |
| Polyclonal | |
| Unconjugated | |
| ACTR10 | |
| Actin related protein 10, Actin related protein 11, ACTR10, ACTR11, ARP11, HARP11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 299121, 55860, 56444 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NZ32, Q9QZB7 | |
| ACTR10 | |
| ACTR10 Fusion Protein Ag13612 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction