missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTR10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56886
This item is not returnable.
View return policy
Description
ACTR10 Polyclonal specifically detects ACTR10 in Human samples. It is validated for Western Blot.
Specifications
| ACTR10 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| actin-related protein 10 homolog (S. cerevisiae), Actin-related protein 11, ACTR11actin-related protein 10, Arp11, HARP11 | |
| Rabbit | |
| 46 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Chicken: 100%; Canine: 100%; Human: 100%; Rat: 100%; Xenopus: 92%; Guinea pig: 92%; Zebrafish: 91%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9NZ32 | |
| ACTR10 | |
| Synthetic peptides corresponding to ACTR10(actin-related protein 10 homolog (S. cerevisiae)) The peptide sequence was selected from the middle region of ACTR10 (NP_060947). Peptide sequence SLIQCPIDTRKQLAENLVVIGGTSMLPGFLHRLLAEIRYLVEKPKYKKAL. | |
| Affinity purified | |
| RUO | |
| 55860 | |
| Centrifuge vial prior to reconstitution. Add 50μL distilled water to a final antibody concentration of 1mg/mL. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction