missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTN4 Mouse anti-Human, Mouse, Rat, Clone: 1E12B8, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ACTN4 |
|---|---|
| Clone | 1E12B8 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16828685
|
Proteintech
66628-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16818685
|
Proteintech
66628-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody recognizes 105 kDa of endogenous ACTN4 protein in various lysates.
Alpha actinins belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments. This gene encodes a nonmuscle, alpha actinin isoform which is concentrated in the cytoplasm, and thought to be involved in metastatic processes. Mutations in this gene have been associated with focal and segmental glomerulosclerosis.Specifications
| ACTN4 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| ACTININ 4, actinin, alpha 4, ACTN4, Alpha actinin 4, F actin cross linking protein, FSGS, FSGS1, Non muscle alpha actinin 4 | |
| Actn4 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1E12B8 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Rat, Mouse, Human | |
| O43707, P57780, Q9QXQ0 | |
| 60595, 63836, 81 | |
| ACTN4 Fusion Protein Ag13549 | |
| Primary | |
| -20°C | |
| ACTN4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title