missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTN4 Mouse anti-Human, Mouse, Rat, Clone: 1E12B8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66628-1-IG-20UL
This item is not returnable.
View return policy
Description
This antibody recognizes 105 kDa of endogenous ACTN4 protein in various lysates.
Alpha actinins belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments. This gene encodes a nonmuscle, alpha actinin isoform which is concentrated in the cytoplasm, and thought to be involved in metastatic processes. Mutations in this gene have been associated with focal and segmental glomerulosclerosis.Specifications
| ACTN4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O43707, P57780, Q9QXQ0 | |
| Actn4 | |
| ACTN4 Fusion Protein Ag13549 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG1 |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1E12B8 | |
| Unconjugated | |
| ACTN4 | |
| ACTININ 4, actinin, alpha 4, ACTN4, Alpha actinin 4, F actin cross linking protein, FSGS, FSGS1, Non muscle alpha actinin 4 | |
| Mouse | |
| Protein G | |
| RUO | |
| 60595, 63836, 81 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur