missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ACTN2 |
|---|---|
| Concentration | 0.19 mg/mL |
| Applications | Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16892754
|
Proteintech
14221-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16882754
|
Proteintech
14221-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alpha actinins belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments.Specifications
| ACTN2 | |
| Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| P35609, Q9JI91 | |
| 11472, 291245, 88 | |
| ACTN2 Fusion Protein Ag5459 | |
| Primary | |
| -20°C | |
| ACTN2 |
| 0.19 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| actinin, alpha 2, ACTN2, Alpha actinin 2, CMD1AA, F actin cross linking protein | |
| Actn2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title