missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activin RIIB Mouse anti-Human, Clone: 6L11, Novus Biologicals™
Mouse Monoclonal Antibody
£193.00
Specifications
| Antigen | Activin RIIB |
|---|---|
| Clone | 6L11 |
| Concentration | LYOPH |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Description
Activin RIIB Monoclonal specifically detects Activin RIIB in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Activin RIIB | |
| LYOPH | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Protein Kinase | |
| 93 | |
| recombinant human Activin receptor IIB extracellular domain | |
| Primary | |
| Reconstitute the antibody with 500 ul sterile PBS and the final concentration is 200 ug/ml. |
| 6L11 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ACVR2B | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title