missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activin RIIB Mouse anti-Human, Clone: 6L11, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36066-0.02MG
This item is not returnable.
View return policy
Description
Activin RIIB Monoclonal specifically detects Activin RIIB in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Activin RIIB | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| activin A receptor, type IIB, Activin receptor type IIB, activin receptor type-2B, ActR-IIB, ACTRIIB, EC 2.7.11, EC 2.7.11.30, MGC116908 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reconstitute the antibody with 500 ul sterile PBS and the final concentration is 200 ug/ml. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 6L11 | |
| Unconjugated | |
| PBS with No Preservative | |
| ACVR2B | |
| recombinant human Activin receptor IIB extracellular domain | |
| 0.02 mg | |
| Protein Kinase | |
| 93 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction