missing translation for 'onlineSavingsMsg'
Learn More

Activated Charcoal, Untreated, 20 - 60 Mesh, Honeywell Fluka™

Product Code. 15647740 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly bottle
Unit Size:
2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15647740 2.5 kg Poly bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15647740 Supplier Honeywell-Fluka Supplier No. C30142.5KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Untreated, granular, 20-60 mesh

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-44-0
Color Black
pH 6-9
Mesh Size 20 to 60mesh
Odor Odorless
Packaging Poly bottle
Molecular Formula C
MDL Number MFCD00133992
Quantity 2.5 kg
UN Number NONH for all modes of transport
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES [C]
IUPAC Name carbon
Molecular Weight (g/mol) 12.011
PubChem CID 5462310
ChEBI CHEBI:27594
Formula Weight 12.01g/mol
Physical Form Powder
Chemical Name or Material Activated charcoal
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.