missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activated Charcoal Norit™, SX2, Honeywell™ Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
Unit Size:
250g
Description
- Norit is a registered trademark of Norit N.V.
Specifications
Specifications
| CAS | 7440-44-0 |
| Color | Black |
| pH | 6 - 9 |
| Odor | Odorless |
| Molecular Formula | C |
| MDL Number | MFCD00133992 |
| Quantity | 250 g |
| UN Number | NONH for all modes of transport |
| Beilstein | 4360473 |
| Synonym | graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction