missing translation for 'onlineSavingsMsg'
Learn More

Activated charcoal, NORIT™ ROW 0.8 SUPRA, pellets, Honeywell Fluka™

Product Code. 15673860 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Drum with inliner
Poly Bottle
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15673860 500 g Poly Bottle 500g
15663860 2 kg Drum with inliner 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15673860 Supplier Honeywell-Fluka Supplier No. 329428500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

NORIT® ROW 0.8 SUPRA, pellets

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Melting Point 3550°C
Color Black
pH 6 to 9
Odor Odourless
Packaging Poly Bottle
Quantity 500 g
UN Number NONH for all modes of transport
Formula Weight 12.01g/mol
Chemical Name or Material Activated Charcoal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.