missing translation for 'onlineSavingsMsg'
Learn More

Activated charcoal, NORIT™ ROW 0.8 SUPRA, pellets, Honeywell Fluka™

Product Code. 15663860 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Drum with inliner
Poly Bottle
Unit Size:
2kg
500g
This item is not returnable. View return policy

Product Code. 15663860

Brand: Honeywell-Fluka 3294282KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

NORIT® ROW 0.8 SUPRA, pellets

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Melting Point 3550°C
Color Black
pH 6 to 9
Odor Odourless
Packaging Drum with inliner
Quantity 2 kg
UN Number NONH for all modes of transport
Formula Weight 12.01g/mol
Chemical Name or Material Activated Charcoal
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.