missing translation for 'onlineSavingsMsg'
Learn More

Activated Charcoal, Norit™ PK 3-5, Honeywell Fluka™

Product Code. 15672950 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15672950 250 g Poly Bottle 250g
15662950 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15672950 Supplier Honeywell-Fluka Supplier No. 292591250G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

4-14 mesh, granular, Norit® PK 3-5

  • Norit is a registered trademark of Norit N.V.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Melting Point 3550°C
Color Black
pH 6 to 9
Packaging Poly Bottle
Quantity 250 g
UN Number NONH for all modes of transport
Formula Weight 12.01g/mol
Grade Norit™ PK 3-5
Physical Form Granules
Chemical Name or Material Activated Charcoal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.