missing translation for 'onlineSavingsMsg'
Learn More

Activated Charcoal Norit™, CA1, Honeywell™ Fluka™

Product Code. 15667590 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15667590 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15667590 Supplier Honeywell-Fluka Supplier No. 978761KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Norit® CA1, from wood, chemically activated, powder

  • Norit is a registered trademark of Norit N.V.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-44-0
Color Black
pH 6 - 9
Odor Odorless
Molecular Formula C
MDL Number MFCD00133992
Quantity 1 kg
UN Number UN1362
Beilstein 4360473
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES [C]
IUPAC Name carbon
Molecular Weight (g/mol) 12.011
PubChem CID 5462310
ChEBI CHEBI:27594
Formula Weight 12.01g/mol
Chemical Name or Material Activated Charcoal Norit™, Norit CA1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.