missing translation for 'onlineSavingsMsg'
Learn More

Activated charcoal, iodinated

Product Code. p-3593362 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10751525 250 g Glass bottle 250g
10360254 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10751525 Supplier Thermo Scientific Acros Supplier No. 389472500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7440-44-0 | C | 12.011 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Color Black
Packaging Glass bottle
Quantity 250 g
Merck Index 14, 1807
Solubility Information Solubility in water: insoluble
Formula Weight 12
Percent Purity 5%, exact strength on the certificate of analysis
Physical Form Thin Rods
Chemical Name or Material Activated charcoal
compliance-icons compliance-icons
Product Identifier
  • Activated charcoal, iodinated
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.