missing translation for 'onlineSavingsMsg'
Learn More

Activated Charcoal, -100 particle size (mesh), Honeywell Fluka™

Product Code. 15641540 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
175 g
1 kg
5 kg
Packaging:
Metal Drum
Plastic Bottle
Unit Size:
175g
1kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15641540 5 kg Metal Drum 5kg
15621540 175 g Plastic Bottle 175g
15631540 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15641540 Supplier Honeywell-Fluka Supplier No. 1615515KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

powder, -100 particle size (mesh), decolorizing

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Melting Point 3550°C (literature)
Color Black
Density ca. 350 kg/cu.m
pH 6 to 9
Mesh Size –100 particle size (mesh)
Packaging Metal Drum
Quantity 5 kg
UN Number NONH for all modes of transport
Formula Weight 12.01g/mol
Physical Form Powder
Chemical Name or Material Activated Charcoal
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.