missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin11 Antibody (mAb2345a) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £324.00
Specifications
| Antigen | Actin11 |
|---|---|
| Clone | mAb2345a |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18240960
|
Novus Biologicals
NBP2-53120 |
0.1 mg |
£324.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603167
|
Novus Biologicals
NBP2-53120-0.025mg |
0.025 mg |
£150.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Actin11 Monoclonal specifically detects Actin11 in Plant samples. It is validated for Western Blot, Immunofluorescence.Specifications
| Actin11 | |
| 1.0 mg/mL | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cellular Signaling | |
| PBS with 0.02% Sodium Azide | |
| Purified Arabidopsis Actin11 | |
| Primary | |
| This mouse monoclonal antibody was generated against plant actin ACT11 and recognizes subclasses 2 and 3 of vegetative and subclasses 4 and 5 of pollen specific plant actins (arabidopsis ACT1, 3, 4, 7, 11, 12). |
| mAb2345a | |
| Western Blot 1:100 - 1:2000, Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Plant | |
| ACT11, Actin-11 | |
| IgG1 | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title