missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin11 Antibody (mAb2345a) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53120-0.025mg
This item is not returnable.
View return policy
Description
Actin11 Monoclonal specifically detects Actin11 in Plant samples. It is validated for Western Blot, Immunofluorescence.
Specifications
| Actin11 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunofluorescence 1:10 - 1:500 | |
| ACT11, Actin-11 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| This mouse monoclonal antibody was generated against plant actin ACT11 and recognizes subclasses 2 and 3 of vegetative and subclasses 4 and 5 of pollen specific plant actins (arabidopsis ACT1, 3, 4, 7, 11, 12). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, Immunofluorescence | |
| mAb2345a | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| TLR4 | |
| Purified Arabidopsis Actin11 | |
| 0.025 mg | |
| Apoptosis, Cancer, Cellular Signaling | |
| 7099 | |
| Plant | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction