Learn More
Actin1 Antibody (mAb45a) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-53123
 View more versions of this product
Product Code. 18260410
  • £324.00 / 0.10mg
Description
Actin1 Monoclonal specifically detects Actin1 in Plant samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Actin1
Monoclonal
1.0 mg/mL
Western Blot 1:100 - 1:2000
AAC1, Actin-1
Purified Arabidopsis Actin1
0.1 mg
Apoptosis, Cancer, Cellular Signaling
Recognizes subclasses 4 and 5 of vegetative plant actins (arabidopsis ACT1, 3, 4, 12).
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot
mAb45a
Unconjugated
PBS with 0.02% Sodium Azide
Mouse
Protein A purified
RUO
Primary
Plant
Purified