missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin1 Antibody (mAb45a) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £324.00
Specifications
| Antigen | Actin1 |
|---|---|
| Clone | mAb45a |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18260410
|
Novus Biologicals
NBP2-53123 |
0.1 mg |
£324.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639856
|
Novus Biologicals
NBP2-53123-0.025mg |
0.025 mg |
£150.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Actin1 Monoclonal specifically detects Actin1 in Plant samples. It is validated for Western Blot.Specifications
| Actin1 | |
| 1.0 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cellular Signaling | |
| PBS with 0.02% Sodium Azide | |
| Purified Arabidopsis Actin1 | |
| Primary | |
| Recognizes subclasses 4 and 5 of vegetative plant actins (arabidopsis ACT1, 3, 4, 12). |
| mAb45a | |
| Western Blot 1:100 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Plant | |
| AAC1, Actin-1 | |
| IgG1 | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title