missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin1 Antibody (mAb45a) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53123-0.025mg
This item is not returnable.
View return policy
Description
Actin1 Monoclonal specifically detects Actin1 in Plant samples. It is validated for Western Blot.
Specifications
| Actin1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000 | |
| AAC1, Actin-1 | |
| Purified Arabidopsis Actin1 | |
| 0.025 mg | |
| Apoptosis, Cancer, Cellular Signaling | |
| Recognizes subclasses 4 and 5 of vegetative plant actins (arabidopsis ACT1, 3, 4, 12). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| mAb45a | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Plant | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction