missing translation for 'onlineSavingsMsg'
Learn More

ACTH Mouse, Clone: CLIP/1449, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 5443-MSM9-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15940678

  • £395.00 / 100µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

ACTH
Cytoplasmic
CLIP/1449
Unconjugated
NP_001306133
Mouse
Normal pituitary gland or pituitary tumor
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
POMC
Synthetic peptide corresponding to aa25-39 of human ACTH (NGAEDESAEAFPLEF)
Protein A/G Purified
RUO
5443
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers