missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTH Mouse, Clone: CLIP/1449, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | ACTH |
---|---|
Clone | CLIP/1449 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15940678
|
EnquireBio
5443-MSM9-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930678
|
EnquireBio
5443-MSM9-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
ACTH | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
5443 | |
Synthetic peptide corresponding to aa25-39 of human ACTH (NGAEDESAEAFPLEF) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
Normal pituitary gland or pituitary tumor |
CLIP/1449 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_001306133 | |
POMC | |
IgG1 | |
Protein A/G Purified | |
Cytoplasmic |