missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTH Mouse, Clone: CLIP/1449, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 5443-MSM9-P0
Additional Details : Weight : 0.01000kg
Specifications
ACTH | |
Cytoplasmic | |
CLIP/1449 | |
Unconjugated | |
NP_001306133 | |
Mouse | |
Normal pituitary gland or pituitary tumor | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
POMC | |
Synthetic peptide corresponding to aa25-39 of human ACTH (NGAEDESAEAFPLEF) | |
Protein A/G Purified | |
RUO | |
5443 | |
IgG1 |