missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACSS2 Recombinant Rabbit Monoclonal Antibody (23GB1705), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | ACSS2 |
|---|---|
| Clone | 23GB1705 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088304
![]() |
Invitrogen™
MA554144 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088312
|
Invitrogen™
MA554145 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACSS2 Recombinant Monoclonal antibody specifically detects ACSS2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| ACSS2 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q9NR19, Q9QXG4 | |
| 311569, 55902, 60525 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1705 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1110017C11Rik; ACAS; Acas1; Acas2; ACECS; AceCS1; acetate thiokinase; acetate-CoA ligase; acetate--CoA ligase; Acetyl-CoA synthetase; Acetyl-CoA synthetase 1; acetyl-Coenzyme A synthetase 1 (AMP forming); acetyl-Coenzyme A synthetase 2 (ADP forming); acetyl-Coenzyme A synthetase 2 (AMP forming); acetyl-coenzyme A synthetase, cytoplasmic; ACS; Acs1; ACSA; ACSS2; Acyl-activating enzyme; acyl-CoA synthetase short-chain family member 2; cytoplasmic acetyl-coenzyme A synthetase; dJ1161H23.1; Propionate--CoA ligase | |
| A synthesized peptide derived from human ACSS2 (100-125AA). | |
| Primary | |
| -20°C | |
| Acss2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title