missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACSS2 Recombinant Rabbit Monoclonal Antibody (23GB1705), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554145
This item is not returnable.
View return policy
Description
This gene encodes a protein with a sterol sensing domain (SSD) and seven WD domains. In the presence of cholesterol, this protein binds to sterol regulatory element binding proteins (SREBPs) and mediates their transport from the ER to the Golgi. The SREBPs are then proteolytically cleaved and regulate sterol biosynthesis.
Specifications
| ACSS2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Acss2 | |
| 1110017C11Rik; ACAS; Acas1; Acas2; ACECS; AceCS1; acetate thiokinase; acetate-CoA ligase; acetate--CoA ligase; Acetyl-CoA synthetase; Acetyl-CoA synthetase 1; acetyl-Coenzyme A synthetase 1 (AMP forming); acetyl-Coenzyme A synthetase 2 (ADP forming); acetyl-Coenzyme A synthetase 2 (AMP forming); acetyl-coenzyme A synthetase, cytoplasmic; ACS; Acs1; ACSA; ACSS2; Acyl-activating enzyme; acyl-CoA synthetase short-chain family member 2; cytoplasmic acetyl-coenzyme A synthetase; dJ1161H23.1; Propionate--CoA ligase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 311569, 55902, 60525 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 23GB1705 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NR19, Q9QXG4 | |
| Acss2 | |
| A synthesized peptide derived from human ACSS2 (100-125AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction