missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSM3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-74081
This item is not returnable.
View return policy
Description
ACSM3 Polyclonal specifically detects ACSM3 in Human samples. It is validated for Western Blot.
Specifications
| ACSM3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| acyl-CoA synthetase medium-chain family member 3Protein SA homolog, acyl-coenzyme A synthetase ACSM3, mitochondrial, Butyrate--CoA ligase 3, Butyryl-coenzyme A synthetase 3, EC 6.2.1, Middle-chain acyl-CoA synthetase 3, SA, SA (rat hypertension-associated) homolog, SA hypertension-associated homolog, SA hypertension-associated homolog (rat), SAH, SAHEC 6.2.1.2 | |
| Rabbit | |
| 66 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Chicken: 78%; Rat: 86%; Zebrafish: 79%; Guinea pig: 79% . | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q53FZ2 | |
| ACSM3 | |
| Synthetic peptides corresponding to thetase medium chain family member 3 Antibody against the C terminal of ACSM3. Immunizing peptide sequence DQEQLIKEIQEHVKKTTAPYKYPRKVEFIQELPKTISGKTKRNELRKKEW. | |
| Affinity purified | |
| RUO | |
| 6296 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction