missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACSL1 Recombinant Rabbit Monoclonal Antibody (7G0L10)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£147.00 - £363.00

Specifications

Antigen ACSL1
Clone 7G0L10
Concentration 1.49 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30283149
View Documents
Invitrogen™
MA558013
20 μL
£147.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30283204
GREENER_CHOICE
View Documents
Invitrogen™
MA558014
100 μL
£363.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Immunogen sequence: ATRPKPLKPP CDLSMQSVEV AGSGGARRSA LLDSDEPLVY FYDDVTTLYE GFQRGIQVSN NGPCLGSRKP DQPYEWLSYK QVAELSECIG SALIQKGFKT.

The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation.
Specifications

Specifications

ACSL1
1.49 mg/mL
ELISA, Western Blot
Unconjugated
Rabbit
RUO
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P18163, P41216
14081, 25288
A synthetic peptide corresponding to a sequence within amino acids 45-144 of human ACSL1 (NP_001986.2).
Antibody
7G0L10
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Mouse, Rat
ACSL1
Acas; Acas1; ACS; Acs1; ACSL1; Acsl2; Acyl CoA synthetase, long chain; acyl-CoA synthetase 1; acyl-CoA synthetase long chain family member 1; acyl-CoA synthetase long-chain family member 1; Arachidonate--CoA ligase; COAA; FACL1; Facl2; FACS; fatty acid Coenzyme A ligase, long chain 2; fatty-acid-Coenzyme A ligase long chain 2; fatty-acid-Coenzyme A ligase, long-chain 1; fatty-acid-Coenzyme A ligase, long-chain 2; L LACS2; LACS; LACS 1; LACS 2; LACS1; LACS2; lignoceroyl-CoA synthase; long-chain acyl-CoA synthetase 1; Long-chain acyl-CoA synthetase 2; long-chain fatty acid-CoA ligase 2; long-chain fatty-acid-coenzyme A ligase 1; long-chain-fatty-acid--CoA ligase 1; Long-chain-fatty-acid--CoA ligase, liver isozyme; OTTHUMP00000218457; OTTHUMP00000218460; OTTHUMP00000218462; OTTHUMP00000218465; palmitoyl-CoA ligase; Palmitoyl-CoA ligase 1; palmitoyl-CoA ligase 2; paltimoyl-CoA ligase 1; Phytanate--CoA ligase
ACSL1
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.