missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACSL1 Recombinant Rabbit Monoclonal Antibody (7G0L10)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558013
This item is not returnable.
View return policy
Description
Immunogen sequence: ATRPKPLKPP CDLSMQSVEV AGSGGARRSA LLDSDEPLVY FYDDVTTLYE GFQRGIQVSN NGPCLGSRKP DQPYEWLSYK QVAELSECIG SALIQKGFKT.
The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation.
Specifications
| ACSL1 | |
| Recombinant Monoclonal | |
| 1.49 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P18163, P41216 | |
| ACSL1 | |
| A synthetic peptide corresponding to a sequence within amino acids 45-144 of human ACSL1 (NP_001986.2). | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 7G0L10 | |
| Unconjugated | |
| ACSL1 | |
| Acas; Acas1; ACS; Acs1; ACSL1; Acsl2; Acyl CoA synthetase, long chain; acyl-CoA synthetase 1; acyl-CoA synthetase long chain family member 1; acyl-CoA synthetase long-chain family member 1; Arachidonate--CoA ligase; COAA; FACL1; Facl2; FACS; fatty acid Coenzyme A ligase, long chain 2; fatty-acid-Coenzyme A ligase long chain 2; fatty-acid-Coenzyme A ligase, long-chain 1; fatty-acid-Coenzyme A ligase, long-chain 2; L LACS2; LACS; LACS 1; LACS 2; LACS1; LACS2; lignoceroyl-CoA synthase; long-chain acyl-CoA synthetase 1; Long-chain acyl-CoA synthetase 2; long-chain fatty acid-CoA ligase 2; long-chain fatty-acid-coenzyme A ligase 1; long-chain-fatty-acid--CoA ligase 1; Long-chain-fatty-acid--CoA ligase, liver isozyme; OTTHUMP00000218457; OTTHUMP00000218460; OTTHUMP00000218462; OTTHUMP00000218465; palmitoyl-CoA ligase; Palmitoyl-CoA ligase 1; palmitoyl-CoA ligase 2; paltimoyl-CoA ligase 1; Phytanate--CoA ligase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 14081, 25288 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction