missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) AcroPrep™ 384-well Filters Plates, 100 μL
AcroPrep™ 384-well Filter Plates are ideal for use in high volume, high throughput sample prep applications.
£458.00 - £785.00
Specifications
Material | Polypropylene |
---|---|
Sterility | Non-sterile |
Quantity | 10/Pk. |
Volume (Metric) | 80 μL |
Product Code | Brand | Pore Size | Material (Membrane) | Type | MWCO | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Material (Membrane) | Type | MWCO | Price | Quantity & Availability | ||
17144471
|
Cytiva (formerly Pall Lab)
5078 |
Omega (Modified PES) | Short Tips | 30 kDa |
£755.00
Pack of 10 |
Estimated Shipment: 17-12-2024
Log in to see stock. |
|||
17184471
|
Cytiva (formerly Pall Lab)
5084 |
1.2 μm | Supor (PES) | Long Tips |
£555.00
Pack of 10 |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||
17194471
|
Cytiva (formerly Pall Lab)
5085 |
1.2 μm | Supor (PES) | Short Tips |
£583.08
Pack of 10 |
Estimated Shipment: 17-12-2024
Log in to see stock. |
|||
17104471
|
Cytiva (formerly Pall Lab)
5072 |
1.0 μm | Borosilicate Glass Fiber | Long Tips | |||||
17104481
|
Cytiva (formerly Pall Lab)
5086 |
0.2 μm | Supor (PES) | Short Tips | |||||
17114471
|
Cytiva (formerly Pall Lab)
5073W |
1.0 μm | Borosilicate Glass Fiber | Short Tips |
£458.00
Pack of 10 |
Estimated Shipment: 17-12-2024
Log in to see stock. |
|||
17124471
|
Cytiva (formerly Pall Lab)
5076 |
Omega (Modified PES) | Short Tips | 10 kDa |
£755.00
Pack of 10 |
Estimated Shipment: 17-12-2024
Log in to see stock. |
|||
17134471
|
Cytiva (formerly Pall Lab)
5077 |
Omega (Modified PES) | Long Tips | 10 kDa |
£755.00
Pack of 10 |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||
17154471
|
Cytiva (formerly Pall Lab)
5079 |
Omega (Modified PES) | Long Tips | 30 kDa |
£755.00
Pack of 10 |
Estimated Shipment: 17-12-2024
Log in to see stock. |
|||
17164471
|
Cytiva (formerly Pall Lab)
5080 |
Omega (Modified PES) | Short Tips | 100 kDa |
£755.00
Pack of 10 |
Estimated Shipment: 12-11-2024
Log in to see stock. |
|||
17174471
|
Cytiva (formerly Pall Lab)
5081 |
Omega (Modified PES) | Long Tips | 100 kDa |
£785.00
Pack of 10 |
Estimated Shipment: 17-12-2024
Log in to see stock. |
|||
Description
- Plates are constructed from chemically-resistant, biologically-inert polypropylene with a clear polystyrene lid
- This product is for research applications only
Specifications
Polypropylene | |
Non-sterile | |
10/Pk. | |
80 μL |