Learn More
Cytiva (formerly Pall Lab) AcroPrep™ 384-well Filters Plates, 100 μL
AcroPrep™ 384-well Filter Plates are ideal for use in high volume, high throughput sample prep applications.
Brand:  Cytiva (formerly Pall Lab) 5073W
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 17114471
  • £458.00 / Pack of 10
Estimated Shipment: 11-09-2024
to see stock.
Add to basket
Description
Description
  • Plates are constructed from chemically-resistant, biologically-inert polypropylene with a clear polystyrene lid
  • This product is for research applications only
Specifications
Specifications
White
Borosilicate Glass Fiber
Non-sterile
Short Tips
Polypropylene
1.0 μm
10/Pk.
80 μL