missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Acrodisc™ PSF Syringe Filters with PVDF Membrane

Syringe Filter with hydrophilic membrane compatible with a wide variety of solvents.

Brand:  Cytiva (formerly Pall Lab) AP-4408

215.65 GBP valid until 2024-05-06
Use promo code "21711" to get your promotional price.



Additional Details : Weight : 1.10200kg

 View more versions of this product

Product Code. 17144671

  • £227.00 / Pack of 50
Estimated Shipment: 05-09-2024
to see stock.
Add to basket

Description

Description

  • Useful for a wide range of applications, including aggressive and non-aggressive solvent-based mobile phase
  • For research use only
Specifications

Specifications

PVDF
Non-sterile
<100 μL
200/Cs.
82°C
0.45 μm
3.9 cm2
Polypropylene
50/Pk.
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers