Learn More
Cytiva (formerly Pall Lab) Acrodisc™ PSF Syringe Filters with PVDF Membrane
Syringe Filter with hydrophilic membrane compatible with a wide variety of solvents.
£132.00 - £2880.00
Specifications
Sterility | Non-sterile |
---|---|
Filtration Area | 3.9 cm2 |
Housing Material | Polypropylene |
Temperature (Metric) Max | 82°C |
Diameter (Metric) | 25 mm |
Product Code | Brand | Pore Size | No. per Pack | Material (Membrane) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | No. per Pack | Material (Membrane) | Price | Quantity & Availability | |||
17144671
|
Cytiva (formerly Pall Lab)
AP-4408 |
0.45 μm | 50/Pk. | PVDF |
£227.00
Pack of 50 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||
17194671
|
Cytiva (formerly Pall Lab)
AP-4500 |
0.45 μm | 1000/Pk. | PVDF |
£2255.00
Pack of 1000 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
17184691
|
Cytiva (formerly Pall Lab)
AP-4793 |
0.2 μm | 50/Pk. | Glass Fiber, PVDF |
£151.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
17114671
|
Cytiva (formerly Pall Lab)
AP-4308 |
0.45 μm | 1000/Pk. | Glass Fiber, PVDF |
£2775.00
Pack of 1000 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
17104701
|
Cytiva (formerly Pall Lab)
AP-4796 |
0.2 μm | 50/Pk. | PVDF |
£132.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
17114701
|
Cytiva (formerly Pall Lab)
AP-4797 |
0.2 μm | 1000/Pk. | PVDF |
£2695.00
Pack of 1000 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
17124671
|
Cytiva (formerly Pall Lab)
AP-4309 |
0.45 μm | 25/AutoPack Tubes | Glass Fiber, PVDF |
£595.00
Pack of 200 |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||
17134671
|
Cytiva (formerly Pall Lab)
AP-4310 |
0.45 μm | 50/Pk. | Glass Fiber, PVDF |
£139.00
Pack of 50 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
17174691
|
Cytiva (formerly Pall Lab)
AP-4792 |
0.2 μm | 25/AutoPack Tubes | Glass Fiber, PVDF |
£665.00
Pack of 200 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||
17194691
|
Cytiva (formerly Pall Lab)
AP-4794 |
0.2 μm | 1000/Pk. | Glass Fiber, PVDF |
£2880.00
Pack of 1000 |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||
Description
- Useful for a wide range of applications, including aggressive and non-aggressive solvent-based mobile phase
- For research use only
Specifications
Non-sterile | |
Polypropylene | |
25 mm |
3.9 cm2 | |
82°C |