missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Acrodisc™ PSF Syringe Filters with PVDF Membrane

Syringe Filter with hydrophilic membrane compatible with a wide variety of solvents.

Brand:  Cytiva (formerly Pall Lab) AP-4310

Additional Details : Weight : 0.93400kg

 View more versions of this product

Product Code. 17134671

  • £139.00 / Pack of 50
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

Description

Description

  • Useful for a wide range of applications, including aggressive and non-aggressive solvent-based mobile phase
  • For research use only
Specifications

Specifications

Glass Fiber, PVDF
Non-sterile
<125 μL
200/Cs.
82°C
0.45 μm
3.9 cm2
Polypropylene
50/Pk.
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers