missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Acrodisc™ PSF Syringe Filters with PVDF Membrane
Syringe Filter with hydrophilic membrane compatible with a wide variety of solvents.
Brand: Cytiva (formerly Pall Lab) AP-4796
128.36 GBP valid until 2024-04-30
Use promo code "21637" to get your promotional price.
Additional Details : Weight : 1.11100kg
Description
- Useful for a wide range of applications, including aggressive and non-aggressive solvent-based mobile phase
- For research use only
Specifications
PVDF | |
Non-sterile | |
<100 μL | |
200/Cs. | |
82°C |
0.2 μm | |
3.9 cm2 | |
Polypropylene | |
50/Pk. | |
25 mm |